Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193509.1 | NP_001180438.1 | 3 - 4 | 418 | 89 |
NM_017735.4 | NP_060205.3 | 3 - 4 | 665 | 89 | |
XM_005264416.1 | XP_005264473.1 | 3 - 4 | 652 | 89 | |
XM_011532958.1 | XP_011531260.1 | 3 - 4 | 652 | 89 | |
GenBank mRNA | AK027570.1 | - | 3 - 4 | 574 | 89 |
AK299196.1 | - | 3 - 4 | 407 | 89 | |
BC001248.1 | - | 2 - 3 | 198 | 89 | |
BC063791.1 | - | 3 - 4 | 623 | 89 |