Assay Design
Gene Transcripts
- Gene Symbol
- MAP4K4
- Entrez Gene ID
-
9448
- Gene Name
- mitogen-activated protein kinase kinase kinase kinase 4
- Gene Aliases
-
FLH21957, HEL-S-31, HGK, MEKKK4, NIK
- Chromosome Location
-
Chr.2: 101697703 - 101894690 on Build GRCh38
View Full Product Details