Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127441.1 | NP_001120913.1 | 7 - 8 | 1048 | 123 |
NM_001127442.1 | NP_001120914.1 | 5 - 6 | 937 | 123 | |
NM_001318223.1 | NP_001305152.1 | 5 - 6 | 937 | 123 | |
NM_080385.4 | NP_525124.3 | 6 - 7 | 995 | 123 | |
XM_005250710.1 | XP_005250767.1 | 6 - 7 | 976 | 123 | |
XM_005250712.1 | XP_005250769.1 | 5 - 6 | 890 | 123 | |
XM_011516698.1 | XP_011515000.1 | 5 - 6 | 1613 | 123 | |
XM_011516701.1 | XP_011515003.1 | 5 - 6 | 1530 | 123 | |
XM_011516702.1 | XP_011515004.1 | 5 - 6 | 1516 | 123 | |
XM_011516703.1 | XP_011515005.1 | 5 - 6 | 1509 | 123 | |
XM_011516704.1 | XP_011515006.1 | 5 - 6 | 1627 | 123 | |
XM_011516705.1 | XP_011515007.1 | 5 - 6 | 1627 | 123 | |
GenBank mRNA | AF384667.1 | - | 4 - 5 | 428 | 123 |
AK093288.1 | - | 5 - 6 | 650 | 123 | |
AK131191.1 | - | 5 - 6 | 873 | 123 | |
AK315256.1 | - | 5 - 6 | 911 | 123 | |
AY155229.1 | - | 6 - 7 | 948 | 123 | |
AY155230.1 | - | 7 - 8 | 1018 | 123 | |
BC039362.1 | - | 7 - 8 | 1022 | 123 | |
BC042996.1 | - | 5 - 6 | 1564 | 123 |