Assay Design



Gene Transcripts

Gene Symbol
CES5A
Entrez Gene ID
221223
Gene Name
carboxylesterase 5A
Gene Aliases
CAUXIN, CES4C1, CES5, CES7, HEL126
Chromosome Location
Chr.16: 55846154 - 55956031 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145024.2 NP_659461.1 10 - 11 1426 70
GenBank mRNA AK056109.1 - 10 - 11 1426 70
BC069548.1 - 10 - 11 1396 70
EU794678.1 - 10 - 11 1426 70

View Full Product Details