Assay Design
Gene Transcripts
- Gene Symbol
- KCTD4
- Entrez Gene ID
-
386618
- Gene Name
- potassium channel tetramerization domain containing 4
- Gene Aliases
-
bA321C24.3
- Chromosome Location
-
Chr.13: 45192853 - 45201040 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_198404.2 |
NP_940686.2 |
1 - 2 |
217 |
79 |
GenBank mRNA |
DA683655.1 |
- |
1 - 2 |
219 |
79 |
View Full Product Details