Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099222.1 | NP_001092692.1 | 11 - 12 | 1099 | 114 |
NM_001099223.1 | NP_001092693.1 | 11 - 12 | 1055 | 114 | |
NM_001099224.1 | NP_001092694.1 | 11 - 12 | 1091 | 114 | |
NM_025103.2 | NP_079379.2 | 11 - 12 | 1091 | 114 | |
XM_011518035.2 | XP_011516337.1 | 11 - 12 | 1091 | 114 | |
XM_011518036.2 | XP_011516338.1 | 4 - 5 | 801 | 114 | |
XM_017015163.1 | XP_016870652.1 | 4 - 5 | 606 | 114 | |
XM_017015164.1 | XP_016870653.1 | 4 - 5 | 2047 | 114 | |
GenBank mRNA | AK023707.1 | - | 11 - 12 | 1052 | 114 |
AK026274.1 | - | 1 - 2 | 81 | 114 | |
AK225570.1 | - | 11 - 12 | 1052 | 114 | |
AY040325.1 | - | 11 - 12 | 1020 | 114 | |
BC023634.2 | - | 11 - 12 | 1036 | 114 | |
BC056875.1 | - | 9 - 10 | 799 | 114 | |
BC107742.1 | - | 11 - 12 | 989 | 114 | |
BC113524.1 | - | 10 - 11 | 864 | 114 | |
BC113526.1 | - | 10 - 11 | 865 | 114 |