Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032149.2 | NP_115525.2 | 1 - 2 | 268 | 85 |
XM_011532315.1 | XP_011530617.1 | 1 - 2 | 248 | 85 | |
GenBank mRNA | AK093085.1 | - | 1 - 2 | 260 | 85 |
AK223345.1 | - | 1 - 2 | 255 | 85 | |
AK314711.1 | - | 1 - 2 | 251 | 85 | |
AL136838.1 | - | 1 - 2 | 229 | 85 | |
DB051883.1 | - | 1 - 2 | 268 | 85 |