Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011511416.2 | XP_011509718.1 | 18 - 19 | 1891 | 80 |
XM_011511419.2 | XP_011509721.1 | 17 - 18 | 1801 | 80 | |
XM_011511423.1 | XP_011509725.1 | 17 - 18 | 1820 | 80 | |
GenBank mRNA | AK001904.1 | - | 12 - 13 | 1150 | 80 |
BC131514.1 | - | 11 - 12 | 1057 | 80 |