Assay Design



Gene Transcripts

Gene Symbol
ACOXL
Entrez Gene ID
55289
Gene Name
acyl-CoA oxidase-like
Gene Aliases
-
Chromosome Location
Chr.2: 110732539 - 111118571 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011511404.2 XP_011509706.1 12 - 13 1315 103
XM_011511405.2 XP_011509707.1 12 - 13 1333 103
XM_011511406.2 XP_011509708.1 12 - 13 1306 103
XM_011511407.2 XP_011509709.1 11 - 12 1066 103
XM_011511409.1 XP_011509711.1 12 - 13 1315 103
XM_011511411.2 XP_011509713.1 11 - 12 1198 103
XM_011511413.1 XP_011509715.1 12 - 13 1315 103
XM_011511414.2 XP_011509716.1 12 - 13 1315 103
XM_011511415.2 XP_011509717.1 12 - 13 1315 103
XM_011511416.2 XP_011509718.1 12 - 13 1315 103
XM_011511420.2 XP_011509722.1 12 - 13 1315 103
XM_011511421.2 XP_011509723.1 12 - 13 1315 103
XM_011511422.1 XP_011509724.1 12 - 13 1315 103
XM_011511423.1 XP_011509725.1 12 - 13 1315 103
XM_011511427.2 XP_011509729.1 12 - 13 1315 103
XM_011511428.2 XP_011509730.1 12 - 13 1315 103
XM_011511429.2 XP_011509731.1 12 - 13 1315 103
XM_011511432.2 XP_011509734.1 12 - 13 1143 103
XM_011511434.2 XP_011509736.1 3 - 4 448 103
XM_017004431.1 XP_016859920.1 11 - 12 1198 103
XM_017004433.1 XP_016859922.1 4 - 5 390 103
XM_017004434.1 XP_016859923.1 4 - 5 350 103
GenBank mRNA BX282060.1 - 3 - 4 264 103

View Full Product Details