Assay Design



Gene Transcripts

Gene Symbol
FRYL
Entrez Gene ID
285527
Gene Name
FRY like transcription coactivator
Gene Aliases
AF4p12, KIAA0826, MOR2
Chromosome Location
Chr.4: 48497363 - 48780312 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015030.1 NP_055845.1 31 - 32 4199 154
XM_005248090.4 XP_005248147.1 32 - 33 4257 154
XM_005248093.4 XP_005248150.1 6 - 7 884 154
XM_011513680.2 XP_011511982.1 29 - 30 3719 154
XM_011513681.2 XP_011511983.1 29 - 30 3719 154
XM_011513682.2 XP_011511984.1 28 - 29 3674 154
XM_011513683.2 XP_011511985.1 31 - 32 4093 154
XM_011513684.2 XP_011511986.1 6 - 7 890 154
XM_011513685.2 XP_011511987.1 29 - 30 3719 154
XM_017008046.1 XP_016863535.1 32 - 33 4524 154
GenBank mRNA CR627228.1 - 15 - 16 2245 154

View Full Product Details