Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015030.1 | NP_055845.1 | 40 - 41 | 5493 | 83 |
XM_005248090.4 | XP_005248147.1 | 41 - 42 | 5551 | 83 | |
XM_005248093.4 | XP_005248150.1 | 15 - 16 | 2178 | 83 | |
XM_011513680.2 | XP_011511982.1 | 38 - 39 | 5013 | 83 | |
XM_011513681.2 | XP_011511983.1 | 38 - 39 | 5013 | 83 | |
XM_011513682.2 | XP_011511984.1 | 37 - 38 | 4968 | 83 | |
XM_011513683.2 | XP_011511985.1 | 40 - 41 | 5387 | 83 | |
XM_011513684.2 | XP_011511986.1 | 15 - 16 | 2184 | 83 | |
XM_017008046.1 | XP_016863535.1 | 41 - 42 | 5818 | 83 | |
GenBank mRNA | AK127307.1 | - | 2 - 3 | 238 | 83 |
AL833170.1 | - | 7 - 8 | 1014 | 83 |