Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015030.1 | NP_055845.1 | 46 - 47 | 6899 | 101 |
XM_005248090.4 | XP_005248147.1 | 47 - 48 | 6957 | 101 | |
XM_005248093.4 | XP_005248150.1 | 21 - 22 | 3584 | 101 | |
XM_011513680.2 | XP_011511982.1 | 44 - 45 | 6416 | 101 | |
XM_011513681.2 | XP_011511983.1 | 44 - 45 | 6419 | 101 | |
XM_011513682.2 | XP_011511984.1 | 43 - 44 | 6374 | 101 | |
XM_011513683.2 | XP_011511985.1 | 46 - 47 | 6793 | 101 | |
XM_011513684.2 | XP_011511986.1 | 21 - 22 | 3590 | 101 | |
XM_017008046.1 | XP_016863535.1 | 47 - 48 | 7224 | 101 | |
GenBank mRNA | AB020633.1 | - | 4 - 5 | 963 | 101 |
AK127307.1 | - | 8 - 9 | 1644 | 101 | |
AL833170.1 | - | 13 - 14 | 2420 | 101 | |
U80082.1 | - | 5 - 6 | 1707 | 101 |