Assay Design



Gene Transcripts

Gene Symbol
MAP2
Entrez Gene ID
4133
Gene Name
microtubule associated protein 2
Gene Aliases
MAP2A, MAP2B, MAP2C
Chromosome Location
Chr.2: 209424047 - 209734118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002374.3 NP_002365.3 7 - 8 4424 124
XM_005246555.1 XP_005246612.1 8 - 9 4789 124
XM_005246565.1 XP_005246622.1 4 - 5 4078 124
XM_005246566.1 XP_005246623.1 4 - 5 4066 124
XM_006712532.1 XP_006712595.1 8 - 9 4786 124
XM_011511186.1 XP_011509488.1 8 - 9 4696 124
XM_011511187.1 XP_011509489.1 7 - 8 4593 124
XM_011511188.1 XP_011509490.1 7 - 8 4593 124
XM_011511190.1 XP_011509492.1 7 - 8 4593 124
XM_011511191.1 XP_011509493.1 7 - 8 4593 124
XM_011511192.1 XP_011509494.1 7 - 8 4593 124
XM_011511193.1 XP_011509495.1 7 - 8 4593 124
XM_011511195.1 XP_011509497.1 6 - 7 4344 124
XM_011511196.2 XP_011509498.1 7 - 8 4531 124
XM_011511197.2 XP_011509499.1 7 - 8 4528 124
XM_017004111.1 XP_016859600.1 7 - 8 4593 124
XM_017004112.1 XP_016859601.1 7 - 8 4543 124
XM_017004113.1 XP_016859602.1 7 - 8 4435 124
XM_017004114.1 XP_016859603.1 7 - 8 4543 124
XM_017004115.1 XP_016859604.1 6 - 7 4335 124
XM_017004116.1 XP_016859605.1 6 - 7 4335 124
XM_017004117.1 XP_016859606.1 6 - 7 4332 124
XM_017004118.1 XP_016859607.1 7 - 8 4450 124
XM_017004119.1 XP_016859608.1 7 - 8 4543 124
XM_017004120.1 XP_016859609.1 7 - 8 4540 124
XM_017004121.1 XP_016859610.1 7 - 8 4450 124
XM_017004122.1 XP_016859611.1 7 - 8 4447 124
XM_017004123.1 XP_016859612.1 7 - 8 4531 124
XM_017004124.1 XP_016859613.1 7 - 8 4438 124
XM_017004125.1 XP_016859614.1 7 - 8 4531 124
XM_017004126.1 XP_016859615.1 7 - 8 4531 124
XM_017004127.1 XP_016859616.1 7 - 8 4528 124
XM_017004128.1 XP_016859617.1 6 - 7 4332 124
XM_017004129.1 XP_016859618.1 6 - 7 4332 124
XM_017004130.1 XP_016859619.1 4 - 5 4515 124
XM_017004131.1 XP_016859620.1 4 - 5 4078 124
XM_017004132.1 XP_016859621.1 4 - 5 4066 124
XM_017004133.1 XP_016859622.1 4 - 5 4078 124
XM_017004134.1 XP_016859623.1 4 - 5 4066 124
XM_017004135.1 XP_016859624.1 4 - 5 4512 124
XM_017004136.1 XP_016859625.1 4 - 5 4078 124
XM_017004137.1 XP_016859626.1 4 - 5 4066 124
GenBank mRNA AK307603.1 - 8 - 9 3271 124
M25668.1 - 1 - 2 2697 124
U01828.1 - 7 - 8 4424 124
U89330.1 - 4 - 5 4164 124

View Full Product Details