Assay Design



Gene Transcripts

Gene Symbol
EME1
Entrez Gene ID
146956
Gene Name
essential meiotic structure-specific endonuclease 1
Gene Aliases
MMS4L, SLX2A
Chromosome Location
Chr.17: 50373220 - 50381479 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166131.1 NP_001159603.1 5 - 6 1196 113
GenBank mRNA BC016470.2 - 4 - 5 1139 113

View Full Product Details