Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152463.2 | NP_689676.2 | 5 - 6 | 1200 | 74 |
XM_005257081.3 | XP_005257138.1 | 5 - 6 | 725 | 74 | |
XM_011524395.2 | XP_011522697.1 | 5 - 6 | 690 | 74 | |
XM_017024236.1 | XP_016879725.1 | 4 - 5 | 1113 | 74 | |
GenBank mRNA | AK055926.1 | - | 5 - 6 | 1191 | 74 |
AK308275.1 | - | 5 - 6 | 690 | 74 |