Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020236.3 | NP_064621.3 | 6 - 7 | 866 | 184 |
XM_011532198.2 | XP_011530500.1 | 7 - 8 | 918 | 184 | |
XM_011532199.1 | XP_011530501.1 | 7 - 8 | 732 | 184 | |
GenBank mRNA | AB049474.1 | - | 5 - 6 | 640 | 184 |
AF212225.1 | - | 6 - 7 | 770 | 184 | |
AK024791.1 | - | 4 - 5 | 469 | 184 | |
AK315600.1 | - | 6 - 7 | 683 | 184 | |
BC014356.1 | - | 6 - 7 | 690 | 184 | |
BC015109.1 | - | 6 - 7 | 679 | 184 | |
BC017765.1 | - | 6 - 7 | 687 | 184 | |
BC032595.1 | - | 6 - 7 | 677 | 184 | |
BU675164.1 | - | 3 - 4 | 184 | ||
CR457191.1 | - | 5 - 6 | 602 | 184 |