Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152384.2 | NP_689597.1 | 10 - 11 | 957 | 104 |
GenBank mRNA | AK097935.1 | - | 2 - 3 | 670 | 104 |
AK312680.1 | - | 9 - 10 | 874 | 104 | |
AL834305.1 | - | 10 - 11 | 948 | 104 | |
AY604003.1 | - | 10 - 11 | 973 | 104 | |
AY604004.1 | - | 9 - 10 | 910 | 104 | |
BC044593.1 | - | 10 - 11 | 954 | 104 | |
KU178857.1 | - | 10 - 11 | 899 | 104 | |
KU178858.1 | - | 9 - 10 | 836 | 104 |