Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152384.2 | NP_689597.1 | 3 - 4 | 269 | 108 |
GenBank mRNA | AK312680.1 | - | 3 - 4 | 249 | 108 |
AL834305.1 | - | 3 - 4 | 260 | 108 | |
AY604003.1 | - | 3 - 4 | 285 | 108 | |
AY604004.1 | - | 3 - 4 | 285 | 108 | |
BC044593.1 | - | 3 - 4 | 266 | 108 | |
BP311042.1 | - | 3 - 4 | 270 | 108 | |
KU178857.1 | - | 3 - 4 | 211 | 108 | |
KU178858.1 | - | 3 - 4 | 211 | 108 |