Assay Design



Gene Transcripts

Gene Symbol
BBS5
Entrez Gene ID
129880
Gene Name
Bardet-Biedl syndrome 5
Gene Aliases
-
Chromosome Location
Chr.2: 169479496 - 169506655 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152384.2 NP_689597.1 4 - 5 313 157
GenBank mRNA AK312680.1 - 4 - 5 293 157
AL834305.1 - 4 - 5 304 157
AY604003.1 - 4 - 5 329 157
AY604004.1 - 4 - 5 329 157
BC044593.1 - 4 - 5 310 157
BP311042.1 - 4 - 5 314 157
KU178857.1 - 4 - 5 255 157
KU178858.1 - 4 - 5 255 157

View Full Product Details