Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004628.4 | NP_004619.3 | 12 - 13 | 2357 | 80 |
NR_027299.1 | 11 - 12 | 2233 | 80 | ||
GenBank mRNA | AK222844.1 | - | 11 - 12 | 2050 | 80 |
AK289761.1 | - | 12 - 13 | 2286 | 80 | |
AK295711.1 | - | 12 - 13 | 2175 | 80 | |
AK311039.1 | - | 11 - 12 | 2163 | 80 | |
BC016620.1 | - | 12 - 13 | 2268 | 80 | |
D21089.1 | - | 12 - 13 | 2358 | 80 | |
HM113488.1 | - | 12 - 13 | 2257 | 80 | |
KJ534962.1 | - | 12 - 13 | 2253 | 80 | |
KJ535085.1 | - | 10 - 11 | 1969 | 80 | |
X65024.1 | - | 11 - 12 | 2093 | 80 |