Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004628.4 | NP_004619.3 | 15 - 16 | 2703 | 90 |
NR_027299.1 | 14 - 15 | 2579 | 90 | ||
GenBank mRNA | AK222844.1 | - | 14 - 15 | 2396 | 90 |
AK289761.1 | - | 15 - 16 | 2632 | 90 | |
AK295711.1 | - | 15 - 16 | 2521 | 90 | |
AK311039.1 | - | 14 - 15 | 2509 | 90 | |
BC016620.1 | - | 15 - 16 | 2614 | 90 | |
D21089.1 | - | 15 - 16 | 2704 | 90 | |
HM113488.1 | - | 15 - 16 | 2603 | 90 | |
KJ534962.1 | - | 15 - 16 | 2599 | 90 | |
KJ535085.1 | - | 13 - 14 | 2315 | 90 | |
X65024.1 | - | 14 - 15 | 2439 | 90 |