Assay Design
Gene Transcripts
- Gene Symbol
- IL36RN
- Entrez Gene ID
-
26525
- Gene Name
- interleukin 36 receptor antagonist
- Gene Aliases
-
FIL1, FIL1(DELTA), FIL1D, IL-36Ra, IL1F5, IL1HY1, IL1L1, IL1RP3, IL36RA, PSORP, PSORS14
- Chromosome Location
-
Chr.2: 113058638 - 113064744 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_012275.2 |
NP_036407.1 |
1 - 2 |
133 |
82 |
GenBank mRNA |
AF186094.1 |
- |
1 - 2 |
134 |
82 |
View Full Product Details