Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198551.1 | NP_001185480.1 | 5 - 6 | 643 | 70 |
NM_024424.3 | NP_077742.2 | 5 - 6 | 1191 | 70 | |
NM_024426.4 | NP_077744.3 | 5 - 6 | 1191 | 70 | |
GenBank mRNA | AB971668.1 | - | 5 - 6 | 1350 | 70 |
AK291736.1 | - | 5 - 6 | 1191 | 70 | |
BC032861.2 | - | 5 - 6 | 638 | 70 | |
BC046461.1 | - | 5 - 6 | 1196 | 70 | |
BG718348.1 | - | 5 - 6 | 648 | 70 | |
X51630.1 | - | 5 - 6 | 1177 | 70 |