Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128159.2 | NP_001121631.1 | 16 - 17 | 1935 | 63 |
NM_018289.3 | NP_060759.2 | 15 - 16 | 1848 | 63 | |
XM_011523953.2 | XP_011522255.1 | 17 - 18 | 2023 | 63 | |
XM_017024817.1 | XP_016880306.1 | 13 - 14 | 1981 | 63 | |
XM_017024818.1 | XP_016880307.1 | 12 - 13 | 2176 | 63 | |
GenBank mRNA | AF246287.1 | - | 16 - 17 | 1843 | 63 |
AF318335.1 | - | 16 - 17 | 1906 | 63 | |
AK001841.1 | - | 6 - 7 | 829 | 63 | |
AK091388.1 | - | 11 - 12 | 1558 | 63 | |
AK092532.1 | - | 17 - 18 | 2033 | 63 | |
AK290343.1 | - | 15 - 16 | 1828 | 63 | |
AY444797.1 | - | 16 - 17 | 1843 | 63 | |
BC006116.1 | - | 15 - 16 | 1760 | 63 | |
EU021218.1 | - | 16 - 17 | 1788 | 63 | |
EU434649.1 | - | 9 - 10 | 957 | 63 | |
EU434650.1 | - | 15 - 16 | 1701 | 63 |