Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291958.1 | NP_001278887.1 | 25 - 26 | 4822 | 71 |
NM_001301072.1 | NP_001288001.1 | 24 - 25 | 4712 | 71 | |
NM_005922.3 | NP_005913.2 | 24 - 25 | 4724 | 71 | |
NM_006724.3 | NP_006715.2 | 23 - 24 | 4574 | 71 | |
NR_120425.1 | 25 - 26 | 4815 | 71 | ||
XM_005266989.2 | XP_005267046.1 | 23 - 24 | 4766 | 71 | |
XM_017010869.1 | XP_016866358.1 | 25 - 26 | 5148 | 71 | |
GenBank mRNA | AF002715.1 | - | 24 - 25 | 4658 | 71 |
AF116604.1 | - | 12 - 13 | 1390 | 71 | |
AK124430.1 | - | 7 - 8 | 1704 | 71 | |
BC136276.1 | - | 24 - 25 | 4608 | 71 | |
BC143735.1 | - | 24 - 25 | 4605 | 71 | |
BC143743.1 | - | 25 - 26 | 4695 | 71 | |
BC143750.1 | - | 25 - 26 | 4702 | 71 | |
BC146770.1 | - | 23 - 24 | 4574 | 71 | |
D86968.2 | - | 23 - 24 | 4574 | 71 |