Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291958.1 | NP_001278887.1 | 2 - 3 | 549 | 68 |
NM_001301072.1 | NP_001288001.1 | 2 - 3 | 549 | 68 | |
NM_005922.3 | NP_005913.2 | 2 - 3 | 549 | 68 | |
NM_006724.3 | NP_006715.2 | 2 - 3 | 549 | 68 | |
NR_120425.1 | 2 - 3 | 549 | 68 | ||
XM_005266989.2 | XP_005267046.1 | 2 - 3 | 753 | 68 | |
XM_017010869.1 | XP_016866358.1 | 3 - 4 | 973 | 68 | |
GenBank mRNA | AF002715.1 | - | 2 - 3 | 486 | 68 |
AK307866.1 | - | 2 - 3 | 418 | 68 | |
BC136276.1 | - | 2 - 3 | 436 | 68 | |
BC143735.1 | - | 2 - 3 | 433 | 68 | |
BC143743.1 | - | 2 - 3 | 432 | 68 | |
BC143750.1 | - | 2 - 3 | 432 | 68 | |
BC146770.1 | - | 2 - 3 | 549 | 68 | |
D86968.2 | - | 2 - 3 | 549 | 68 |