Assay Design



Gene Transcripts

Gene Symbol
SDCCAG8
Entrez Gene ID
10806
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
Chromosome Location
Chr.1: 243255328 - 243500092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006642.3 NP_006633.1 10 - 11 1389 97
XM_005273013.4 XP_005273070.1 9 - 10 1221 97
XM_005273018.1 XP_005273075.1 9 - 10 1228 97
XM_005273021.4 XP_005273078.1 10 - 11 1161 97
XM_005273022.3 XP_005273079.1 7 - 8 1010 97
XM_006711727.3 XP_006711790.1 10 - 11 1275 97
XM_006711728.3 XP_006711791.1 9 - 10 1146 97
XM_011544021.2 XP_011542323.1 11 - 12 1371 97
XM_011544022.2 XP_011542324.1 11 - 12 1446 97
XM_011544023.2 XP_011542325.1 11 - 12 1371 97
XM_011544024.2 XP_011542326.1 11 - 12 1371 97
XM_011544026.2 XP_011542328.1 11 - 12 1371 97
XM_011544028.2 XP_011542330.1 9 - 10 1146 97
XM_011544030.2 XP_011542332.1 3 - 4 1922 97
XM_017000103.1 XP_016855592.1 11 - 12 1371 97
XM_017000104.1 XP_016855593.1 9 - 10 1221 97
XM_017000105.1 XP_016855594.1 10 - 11 1376 97
XM_017000106.1 XP_016855595.1 10 - 11 1357 97
XM_017000107.1 XP_016855596.1 11 - 12 1273 97
XM_017000108.1 XP_016855597.1 13 - 14 1820 97
GenBank mRNA AF039690.1 - 4 - 5 560 97
AF250731.1 - 10 - 11 1220 97
AK297105.1 - 6 - 7 808 97
AK299212.1 - 10 - 11 1338 97
AK299249.1 - 7 - 8 981 97

View Full Product Details