Assay Design



Gene Transcripts

Gene Symbol
SDCCAG8
Entrez Gene ID
10806
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
Chromosome Location
Chr.1: 243255328 - 243500092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006642.3 NP_006633.1 8 - 9 1088 93
XM_005273013.4 XP_005273070.1 7 - 8 920 93
XM_005273018.1 XP_005273075.1 7 - 8 927 93
XM_005273021.4 XP_005273078.1 8 - 9 860 93
XM_005273023.4 XP_005273080.1 8 - 9 1765 93
XM_006711727.3 XP_006711790.1 8 - 9 974 93
XM_006711728.3 XP_006711791.1 7 - 8 845 93
XM_011544021.2 XP_011542323.1 9 - 10 1070 93
XM_011544022.2 XP_011542324.1 9 - 10 1145 93
XM_011544023.2 XP_011542325.1 9 - 10 1070 93
XM_011544024.2 XP_011542326.1 9 - 10 1070 93
XM_011544026.2 XP_011542328.1 9 - 10 1070 93
XM_011544028.2 XP_011542330.1 7 - 8 845 93
XM_011544029.2 XP_011542331.1 9 - 10 1070 93
XM_017000103.1 XP_016855592.1 9 - 10 1070 93
XM_017000104.1 XP_016855593.1 7 - 8 920 93
XM_017000105.1 XP_016855594.1 8 - 9 1075 93
XM_017000106.1 XP_016855595.1 8 - 9 1056 93
XM_017000107.1 XP_016855596.1 9 - 10 972 93
XM_017000108.1 XP_016855597.1 11 - 12 1519 93
XM_017000109.1 XP_016855598.1 8 - 9 974 93
GenBank mRNA AF039690.1 - 2 - 3 259 93
AF161348.1 - 8 - 9 1065 93
AF250731.1 - 8 - 9 919 93
AK297105.1 - 4 - 5 507 93
AK299212.1 - 8 - 9 1037 93
AK299249.1 - 5 - 6 680 93
BC032454.1 - 8 - 9 1031 93

View Full Product Details