Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270425.1 | NP_001257354.1 | 24 - 25 | 2114 | 154 |
NM_003618.3 | NP_003609.2 | 25 - 26 | 2177 | 154 | |
GenBank mRNA | AK291467.1 | - | 25 - 26 | 2143 | 154 |
AK299889.1 | - | 17 - 18 | 1206 | 154 | |
AK314487.1 | - | 24 - 25 | 1951 | 154 | |
BC071579.1 | - | 24 - 25 | 2106 | 154 |