Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270425.1 | NP_001257354.1 | 26 - 27 | 2294 | 80 |
NM_003618.3 | NP_003609.2 | 27 - 28 | 2357 | 80 | |
GenBank mRNA | AK291467.1 | - | 27 - 28 | 2323 | 80 |
AK299889.1 | - | 19 - 20 | 1386 | 80 | |
AK314487.1 | - | 26 - 27 | 2131 | 80 | |
BC071579.1 | - | 26 - 27 | 2286 | 80 |