Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270425.1 | NP_001257354.1 | 27 - 28 | 2403 | 128 |
NM_003618.3 | NP_003609.2 | 28 - 29 | 2466 | 128 | |
GenBank mRNA | AK291467.1 | - | 28 - 29 | 2432 | 128 |
AK299889.1 | - | 20 - 21 | 1495 | 128 | |
AK314487.1 | - | 27 - 28 | 2240 | 128 | |
BC071579.1 | - | 27 - 28 | 2395 | 128 |