Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270425.1 | NP_001257354.1 | 3 - 4 | 570 | 126 |
NM_003618.3 | NP_003609.2 | 3 - 4 | 570 | 126 | |
GenBank mRNA | AK291467.1 | - | 3 - 4 | 536 | 126 |
AK314487.1 | - | 3 - 4 | 407 | 126 | |
BC071579.1 | - | 3 - 4 | 562 | 126 |