Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270425.1 | NP_001257354.1 | 9 - 10 | 984 | 141 |
NM_003618.3 | NP_003609.2 | 9 - 10 | 984 | 141 | |
GenBank mRNA | AK314487.1 | - | 9 - 10 | 821 | 141 |
BC071579.1 | - | 9 - 10 | 976 | 141 |