Assay Design



Gene Transcripts

Gene Symbol
ITIH5
Entrez Gene ID
80760
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 5
Gene Aliases
ITI-HC5, PP14776
Chromosome Location
Chr.10: 7559270 - 7666998 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001851.2 NP_001001851.1 9 - 10 1522 129
NM_030569.6 NP_085046.5 9 - 10 1522 129
NM_032817.5 NP_116206.4 5 - 6 857 129
XM_011519713.2 XP_011518015.1 10 - 11 1597 129
XM_011519714.2 XP_011518016.1 10 - 11 1597 129
GenBank mRNA AB075833.1 - 6 - 7 1063 129
AK027375.1 - 9 - 10 1459 129
AK027831.1 - 6 - 7 1041 129
AK075381.1 - 9 - 10 1495 129
AL833203.1 - 9 - 10 1367 129
AY238437.1 - 9 - 10 1418 129
AY358426.1 - 9 - 10 1487 129
BC136439.1 - 9 - 10 1495 129
CR627109.1 - 4 - 5 5956 129

View Full Product Details