Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300754.1 | NP_001287683.1 | 5 - 6 | 803 | 72 |
NM_001300756.1 | NP_001287685.1 | 5 - 6 | 1169 | 72 | |
NM_178833.5 | NP_849155.2 | 6 - 7 | 1340 | 72 | |
XM_006714085.2 | XP_006714148.1 | 6 - 7 | 1339 | 72 | |
XM_006714086.3 | XP_006714149.1 | 5 - 6 | 1168 | 72 | |
GenBank mRNA | AK172823.1 | - | 6 - 7 | 1340 | 72 |
AK297010.1 | - | 5 - 6 | 714 | 72 | |
AK299888.1 | - | 5 - 6 | 728 | 72 | |
BC009732.1 | - | 3 - 4 | 408 | 72 | |
BC047447.1 | - | 6 - 7 | 870 | 72 |