Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174087.1 | NP_001167558.1 | 1 - 2 | 161 | 61 |
NM_001174088.1 | NP_001167559.1 | 1 - 2 | 161 | 61 | |
NM_006534.3 | NP_006525.2 | 1 - 2 | 161 | 61 | |
NM_181659.2 | NP_858045.1 | 1 - 2 | 161 | 61 | |
GenBank mRNA | AB209863.1 | - | 1 - 2 | 98 | 61 |
AF012108.1 | - | 1 - 2 | 100 | 61 | |
AF016031.1 | - | 1 - 2 | 91 | 61 | |
AF036892.1 | - | 1 - 2 | 83 | 61 | |
AK289683.1 | - | 1 - 2 | 105 | 61 | |
AK308346.1 | - | 1 - 2 | 105 | 61 | |
BC092516.1 | - | 1 - 2 | 116 | 61 | |
BC119001.1 | - | 1 - 2 | 49 | 61 | |
BC122547.1 | - | 1 - 2 | 48 | 61 | |
BU588121.1 | - | 1 - 2 | 108 | 61 | |
DB091786.1 | - | 1 - 2 | 161 | 61 |