Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174087.1 | NP_001167558.1 | 4 - 5 | 516 | 115 |
NM_001174088.1 | NP_001167559.1 | 4 - 5 | 516 | 115 | |
NM_006534.3 | NP_006525.2 | 4 - 5 | 516 | 115 | |
NM_181659.2 | NP_858045.1 | 4 - 5 | 516 | 115 | |
GenBank mRNA | AB209863.1 | - | 4 - 5 | 453 | 115 |
AF010227.1 | - | 3 - 4 | 340 | 115 | |
AF012108.1 | - | 4 - 5 | 455 | 115 | |
AF016031.1 | - | 4 - 5 | 446 | 115 | |
AF036892.1 | - | 4 - 5 | 438 | 115 | |
AK289683.1 | - | 4 - 5 | 460 | 115 | |
AK308346.1 | - | 4 - 5 | 460 | 115 | |
BC092516.1 | - | 4 - 5 | 471 | 115 | |
BC119001.1 | - | 4 - 5 | 404 | 115 | |
BC122547.1 | - | 4 - 5 | 403 | 115 |