Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019111.4 | NP_061984.2 | 2 - 3 | 434 | 83 |
GenBank mRNA | AK297032.1 | - | 2 - 3 | 397 | 83 |
AK304429.1 | - | 2 - 3 | 280 | 83 | |
AK313123.1 | - | 2 - 3 | 389 | 83 | |
BC032350.1 | - | 2 - 3 | 365 | 83 | |
BC071659.1 | - | 2 - 3 | 373 | 83 | |
BM849755.1 | - | 2 - 3 | 435 | 83 | |
CR457013.1 | - | 2 - 3 | 325 | 83 | |
J00194.1 | - | 2 - 3 | 351 | 83 | |
K01171.1 | - | 2 - 3 | 339 | 83 | |
M60334.1 | - | 2 - 3 | 352 | 83 |