Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134477.2 | NP_001127949.1 | 7 - 8 | 617 | 110 |
NM_001242992.1 | NP_001229921.1 | 6 - 7 | 562 | 110 | |
NM_002582.3 | NP_002573.1 | 7 - 8 | 700 | 110 | |
XM_011522510.2 | XP_011520812.1 | 7 - 8 | 706 | 110 | |
XM_011522511.2 | XP_011520813.1 | 7 - 8 | 705 | 110 | |
XM_011522513.2 | XP_011520815.1 | 7 - 8 | 616 | 110 | |
XM_011522514.1 | XP_011520816.1 | 7 - 8 | 704 | 110 | |
XM_017023258.1 | XP_016878747.1 | 7 - 8 | 704 | 110 | |
XM_017023259.1 | XP_016878748.1 | 4 - 5 | 349 | 110 | |
XM_017023260.1 | XP_016878749.1 | 2 - 3 | 238 | 110 | |
GenBank mRNA | AJ005698.1 | - | 7 - 8 | 611 | 110 |
AK293189.1 | - | 7 - 8 | 609 | 110 | |
AK301648.1 | - | 6 - 7 | 552 | 110 | |
AK315020.1 | - | 7 - 8 | 696 | 110 | |
BC044763.1 | - | 7 - 8 | 638 | 110 | |
BC050029.1 | - | 7 - 8 | 638 | 110 |