Assay Design



Gene Transcripts

Gene Symbol
EPN2
Entrez Gene ID
22905
Gene Name
epsin 2
Gene Aliases
EHB21
Chromosome Location
Chr.17: 19237377 - 19336715 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014964.4 NP_055779.2 3 - 4 1048 93
GenBank mRNA AB028988.1 - 3 - 4 1013 93
AF062085.1 - 1 - 2 600 93
AK001491.1 - 1 - 2 741 93

View Full Product Details