Assay Design



Gene Transcripts

Gene Symbol
EPN2
Entrez Gene ID
22905
Gene Name
epsin 2
Gene Aliases
EHB21
Chromosome Location
Chr.17: 19237377 - 19336715 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102664.1 NP_001096134.1 3 - 4 359 65
NM_014964.4 NP_055779.2 6 - 7 1418 65
NM_148921.3 NP_683723.2 5 - 6 1247 65
GenBank mRNA AB028988.1 - 6 - 7 1383 65
AF062084.1 - 3 - 4 799 65
AF062085.1 - 4 - 5 973 65
AK001491.1 - 4 - 5 1111 65
AK001996.1 - 3 - 4 966 65
AK024115.1 - 5 - 6 1227 65
BC070036.1 - - 1212 65
BC093972.1 - 3 - 4 872 65
BC093974.1 - 3 - 4 872 65
DA220261.1 - 3 - 4 340 65

View Full Product Details