Assay Design



Gene Transcripts

Gene Symbol
THAP6
Entrez Gene ID
152815
Gene Name
THAP domain containing 6
Gene Aliases
-
Chromosome Location
Chr.4: 75513951 - 75547334 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011531667.2 XP_011529969.1 1 - 2 42 75

View Full Product Details