Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037808.2 | NP_001032897.1 | 3 - 4 | 355 | 57 |
NM_001199142.1 | NP_001186071.1 | 3 - 4 | 475 | 57 | |
NM_001267574.2 | NP_001254503.1 | 2 - 3 | 444 | 57 | |
NM_001286478.1 | NP_001273407.1 | 3 - 4 | 306 | 57 | |
NM_003752.4 | NP_003743.1 | 3 - 4 | 306 | 57 | |
XM_017023814.1 | XP_016879303.1 | 3 - 4 | 261 | 57 | |
GenBank mRNA | AK000739.1 | - | 2 - 3 | 336 | 57 |
AK055366.1 | - | 3 - 4 | 249 | 57 | |
AK292155.1 | - | 3 - 4 | 475 | 57 | |
AK293264.1 | - | 3 - 4 | 247 | 57 | |
AK299421.1 | - | 3 - 4 | 249 | 57 | |
AK301183.1 | - | 3 - 4 | 259 | 57 | |
AK301233.1 | - | 3 - 4 | 258 | 57 | |
AK303790.1 | - | 3 - 4 | 249 | 57 | |
AL045958.1 | - | 3 - 4 | 258 | 57 | |
AY762099.1 | - | 3 - 4 | 218 | 57 | |
BC000533.2 | - | 3 - 4 | 236 | 57 | |
BC001571.1 | - | 3 - 4 | 240 | 57 | |
BC071705.1 | - | 3 - 4 | 234 | 57 | |
BC107692.1 | - | 3 - 4 | 266 | 57 | |
BC157842.1 | - | 3 - 4 | 364 | 57 | |
BC157849.1 | - | 2 - 3 | 204 | 57 | |
BE780094.1 | - | 3 - 4 | 284 | 57 | |
BM757468.1 | - | 3 - 4 | 260 | 57 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099661.2 | NP_001093131.1 | 3 - 4 | 307 | 57 |
NM_001317856.1 | NP_001304785.1 | 3 - 4 | 352 | 57 | |
NM_001317857.1 | NP_001304786.1 | 3 - 4 | 303 | 57 | |
XM_017023620.1 | XP_016879109.1 | 3 - 4 | 259 | 57 | |
GenBank mRNA | BQ686089.1 | - | 3 - 4 | 272 | 57 |
DA645577.1 | - | 3 - 4 | 253 | 57 | |
DC410988.1 | - | 3 - 4 | 303 | 57 |