Assay Design



Gene Transcripts

Gene Symbol
COL13A1
Entrez Gene ID
1305
Gene Name
collagen type XIII alpha 1 chain
Gene Aliases
CMS19, COLXIIIA1
Chromosome Location
Chr.10: 69801885 - 69959148 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320951.1 NP_001307880.1 32 - 33 2301 82
NM_080800.3 NP_542990.3 32 - 33 2319 82
NM_080802.3 NP_542992.3 31 - 32 2265 82
NM_080805.3 NP_542995.3 29 - 30 2178 82
XM_011539295.2 XP_011537597.1 33 - 34 2367 82
XM_017015677.1 XP_016871166.1 34 - 35 2412 82
XM_017015679.1 XP_016871168.1 33 - 34 2367 82
XM_017015682.1 XP_016871171.1 32 - 33 2325 82
XM_017015687.1 XP_016871176.1 33 - 34 2367 82
XM_017015688.1 XP_016871177.1 31 - 32 2262 82
XM_017015690.1 XP_016871179.1 29 - 30 2235 82
XM_017015691.1 XP_016871180.1 32 - 33 2304 82
XM_017015692.1 XP_016871181.1 30 - 31 2226 82
XM_017015697.1 XP_016871186.1 31 - 32 1735 82
GenBank mRNA BY796799.2 - 4 - 5 213 82
M15524.1 - 3 - 4 125 82

View Full Product Details