Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006623.3 | NP_006614.2 | 2 - 3 | 513 | 61 |
XM_011541226.1 | XP_011539528.1 | 2 - 3 | 572 | 61 | |
XM_011541227.2 | XP_011539529.1 | 2 - 3 | 277 | 61 | |
XM_011541228.1 | XP_011539530.1 | 2 - 3 | 190 | 61 | |
XM_011541231.2 | XP_011539533.1 | 2 - 3 | 374 | 61 | |
GenBank mRNA | AF171237.1 | - | 2 - 3 | 359 | 61 |
AK093306.1 | - | 1 - 2 | 1691 | 61 | |
AK309662.1 | - | 2 - 3 | 373 | 61 | |
AK310132.1 | - | 2 - 3 | 373 | 61 | |
AK315360.1 | - | 2 - 3 | 434 | 61 | |
BC000303.2 | - | 2 - 3 | 353 | 61 | |
BC001349.2 | - | 2 - 3 | 348 | 61 | |
BC011262.1 | - | 2 - 3 | 422 | 61 | |
BC023235.1 | - | 2 - 3 | 326 | 61 | |
CR456795.1 | - | 2 - 3 | 286 | 61 | |
DA731339.1 | - | 2 - 3 | 594 | 61 | |
FJ224347.1 | - | - | 978 | 61 |