Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322042.1 | NP_001308971.1 | 8 - 9 | 1895 | 136 |
NM_024596.4 | NP_078872.2 | 8 - 9 | 1895 | 136 | |
XM_011534755.2 | XP_011533057.1 | 8 - 9 | 1895 | 136 | |
XM_011534756.2 | XP_011533058.1 | 8 - 9 | 1894 | 136 | |
XM_011534757.2 | XP_011533059.1 | 8 - 9 | 1894 | 136 | |
XM_011534758.2 | XP_011533060.1 | 8 - 9 | 1894 | 136 | |
XM_011534759.2 | XP_011533061.1 | 8 - 9 | 1894 | 136 | |
XM_011534760.2 | XP_011533062.1 | 4 - 5 | 1471 | 136 | |
XM_017013829.1 | XP_016869318.1 | 8 - 9 | 1895 | 136 | |
XM_017013830.1 | XP_016869319.1 | 8 - 9 | 1894 | 136 | |
XM_017013831.1 | XP_016869320.1 | 8 - 9 | 1894 | 136 | |
XM_017013832.1 | XP_016869321.1 | 8 - 9 | 1894 | 136 | |
XM_017013833.1 | XP_016869322.1 | 8 - 9 | 1894 | 136 | |
XM_017013834.1 | XP_016869323.1 | 8 - 9 | 1894 | 136 | |
GenBank mRNA | AK022909.1 | - | 5 - 6 | 1534 | 136 |