Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005777.2 | NP_005768.1 | 3 - 4 | 1566 | 87 |
XM_005264784.1 | XP_005264841.1 | 3 - 4 | 1841 | 87 | |
XM_005264787.2 | XP_005264844.1 | 1 - 2 | 995 | 87 | |
XM_006712916.1 | XP_006712979.1 | 3 - 4 | 1666 | 87 | |
XM_017005496.1 | XP_016860985.1 | 1 - 2 | 995 | 87 | |
XM_017005499.1 | XP_016860988.1 | 3 - 4 | 1841 | 87 | |
XM_017005500.1 | XP_016860989.1 | 3 - 4 | 1841 | 87 | |
XM_017005501.1 | XP_016860990.1 | 3 - 4 | 1841 | 87 | |
GenBank mRNA | AF042857.1 | - | 3 - 4 | 1419 | 87 |
AF069517.1 | - | 3 - 4 | 1522 | 87 | |
AF091264.1 | - | 3 - 4 | 1451 | 87 | |
AK291719.1 | - | 3 - 4 | 1566 | 87 | |
AK303371.1 | - | 3 - 4 | 1459 | 87 | |
U50839.1 | - | 2 - 3 | 1352 | 87 |