Assay Design



Gene Transcripts

Gene Symbol
MELK
Entrez Gene ID
9833
Gene Name
maternal embryonic leucine zipper kinase
Gene Aliases
HPK38
Chromosome Location
Chr.9: 36572862 - 36677683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256685.1 NP_001243614.1 7 - 8 752 77
NM_001256687.1 NP_001243616.1 6 - 7 608 77
NM_001256688.1 NP_001243617.1 5 - 6 539 77
NM_001256689.1 NP_001243618.1 6 - 7 666 77
NM_001256690.1 NP_001243619.1 5 - 6 549 77
NM_001256691.1 NP_001243620.1 5 - 6 522 77
NM_001256692.1 NP_001243621.1 4 - 5 426 77
NM_014791.3 NP_055606.1 7 - 8 752 77
XM_011518076.2 XP_011516378.1 7 - 8 931 77
XM_011518077.1 XP_011516379.1 8 - 9 827 77
XM_011518078.2 XP_011516380.1 8 - 9 1014 77
XM_011518079.1 XP_011516381.1 7 - 8 704 77
XM_011518080.1 XP_011516382.1 7 - 8 719 77
XM_011518081.2 XP_011516383.1 7 - 8 737 77
XM_011518082.2 XP_011516384.1 6 - 7 845 77
XM_011518083.2 XP_011516385.1 7 - 8 928 77
XM_011518084.2 XP_011516386.1 6 - 7 540 77
XM_011518085.1 XP_011516387.1 5 - 6 536 77
XM_011518086.2 XP_011516388.1 7 - 8 719 77
GenBank mRNA AB183427.1 - 6 - 7 617 77
AB183428.1 - 5 - 6 502 77
AK222811.1 - 7 - 8 751 77
AK293284.1 - 5 - 6 504 77
AK293423.1 - 5 - 6 487 77
AK299164.1 - 7 - 8 704 77
AK300761.1 - 6 - 7 573 77
AK300821.1 - 6 - 7 620 77
AK302374.1 - 4 - 5 393 77
BC014039.2 - 7 - 8 706 77
D79997.1 - 7 - 8 738 77

View Full Product Details