Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256685.1 | NP_001243614.1 | 9 - 10 | 919 | 104 |
NM_001256687.1 | NP_001243616.1 | 8 - 9 | 775 | 104 | |
NM_001256688.1 | NP_001243617.1 | 7 - 8 | 706 | 104 | |
NM_001256689.1 | NP_001243618.1 | 8 - 9 | 833 | 104 | |
NM_001256690.1 | NP_001243619.1 | 7 - 8 | 716 | 104 | |
NM_001256691.1 | NP_001243620.1 | 7 - 8 | 689 | 104 | |
NM_001256692.1 | NP_001243621.1 | 6 - 7 | 593 | 104 | |
NM_001256693.1 | NP_001243622.1 | 7 - 8 | 733 | 104 | |
NM_014791.3 | NP_055606.1 | 9 - 10 | 919 | 104 | |
NR_046337.1 | 9 - 10 | 956 | 104 | ||
XM_011518076.2 | XP_011516378.1 | 9 - 10 | 1098 | 104 | |
XM_011518077.1 | XP_011516379.1 | 10 - 11 | 994 | 104 | |
XM_011518078.2 | XP_011516380.1 | 10 - 11 | 1181 | 104 | |
XM_011518079.1 | XP_011516381.1 | 9 - 10 | 871 | 104 | |
XM_011518080.1 | XP_011516382.1 | 9 - 10 | 886 | 104 | |
XM_011518081.2 | XP_011516383.1 | 9 - 10 | 904 | 104 | |
XM_011518082.2 | XP_011516384.1 | 8 - 9 | 1012 | 104 | |
XM_011518083.2 | XP_011516385.1 | 9 - 10 | 1095 | 104 | |
XM_011518084.2 | XP_011516386.1 | 8 - 9 | 707 | 104 | |
XM_011518085.1 | XP_011516387.1 | 7 - 8 | 703 | 104 | |
XM_011518086.2 | XP_011516388.1 | 9 - 10 | 886 | 104 | |
GenBank mRNA | AB183427.1 | - | 8 - 9 | 784 | 104 |
AB183428.1 | - | 7 - 8 | 669 | 104 | |
AK222811.1 | - | 9 - 10 | 918 | 104 | |
AK293284.1 | - | 7 - 8 | 671 | 104 | |
AK293423.1 | - | 7 - 8 | 654 | 104 | |
AK293447.1 | - | 7 - 8 | 673 | 104 | |
AK296820.1 | - | 9 - 10 | 955 | 104 | |
AK299164.1 | - | 9 - 10 | 871 | 104 | |
AK300761.1 | - | 8 - 9 | 740 | 104 | |
AK300821.1 | - | 8 - 9 | 787 | 104 | |
AK301131.1 | - | 7 - 8 | 665 | 104 | |
AK302374.1 | - | 6 - 7 | 560 | 104 | |
AK309470.1 | - | 8 - 9 | 792 | 104 | |
BC014039.2 | - | 9 - 10 | 873 | 104 | |
D79997.1 | - | 9 - 10 | 905 | 104 |