Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126123.3 | NP_001119595.1 | 3 - 4 | 360 | 137 |
NM_001318759.1 | NP_001305688.1 | 4 - 5 | 478 | 137 | |
NM_017512.5 | NP_059982.2 | 4 - 5 | 476 | 137 | |
NM_202758.3 | NP_974487.1 | 4 - 5 | 475 | 137 | |
XM_011525679.2 | XP_011523981.2 | 4 - 5 | 455 | 137 | |
XM_017025833.1 | XP_016881322.1 | 4 - 5 | 462 | 137 | |
XM_017025835.1 | XP_016881324.1 | 3 - 4 | 348 | 137 | |
GenBank mRNA | AK127818.1 | - | 4 - 5 | 574 | 137 |
AK292780.1 | - | 4 - 5 | 444 | 137 | |
AK309035.1 | - | 3 - 4 | 348 | 137 | |
AW001055.1 | - | 3 - 4 | 137 | ||
BC001285.1 | - | 4 - 5 | 476 | 137 | |
BP265528.1 | - | 4 - 5 | 482 | 137 | |
BP271162.1 | - | 4 - 5 | 429 | 137 | |
BQ710874.1 | - | 4 - 5 | 597 | 137 | |
X67098.1 | - | 3 - 4 | 221 | 137 | |
X89602.1 | - | 4 - 5 | 337 | 137 |